accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CONHSOHO

CAS RN 20640-68-0

IUPAC Name: 2-acetamido-3-(phenylsulfanyl)propanoic acid
Synonyms: 2-acetamido-3-(phenylsulfanyl)propanoic acid
Molecular Weight (g/mol): 239.29
Molecular Formula: C11H13NO3S
InChi Key: CICOZWHZVMOPJS-UHFFFAOYNA-N
SMILES: CC(=O)NC(CSC1=CC=CC=C1)C(O)=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results