CAS RN 2068-78-2
MilliporeSigma™ Calbiochem™ Vincristine Sulfate, Apocynaceae sp.
Indole alkaloid that binds to tubulin, inhibiting formation of microtubules.
Thermo Scientific Chemicals Vincristine sulfate, 95%
CAS: 2068-78-2 Molecular Formula: C46H56N4O10·H2SO4 Molecular Weight (g/mol): 923.03 MDL Number: MFCD08706469 InChI Key: AQTQHPDCURKLKT-DJOZZVTQSA-N Synonym: vincristine sulfate,methyl 9r,10s,11r,12r,19r-11-acetyloxy-12-ethyl-4-13s,15r,17s-17-ethyl-17-hydroxy-13-methoxycarbonyl-1,11-diazatetracyclo 13.3.1.0?, 1 2 .0?, 1 ? nonadeca-4 12 ,5,7,9-tetraen-13-yl-8-formyl-10-hydroxy-5-methoxy-8,16-diazapentacyclo 10.6.1.0 1 ,?.0 2 ,?.0 PubChem CID: 126963557 SMILES: CCC1(CC2CC(C3=C(CCN(C2)C1)C4=CC=CC=C4N3)(C5=C(C=C6C(=C5)C78CCN9C7C(C=CC9)(C(C(C8N6C=O)(C(=O)OC)O)OC(=O)C)CC)OC)C(=O)OC)O.OS(=O)(=O)O
Vincristine sulfate, 97+%
CAS: 2068-78-2 Molecular Formula: C46H58N4O14S Molecular Weight (g/mol): 923.044 MDL Number: MFCD00084729 InChI Key: AQTQHPDCURKLKT-DJOZZVTQSA-N PubChem CID: 126963557 SMILES: CCC1(CC2CC(C3=C(CCN(C2)C1)C4=CC=CC=C4N3)(C5=C(C=C6C(=C5)C78CCN9C7C(C=CC9)(C(C(C8N6C=O)(C(=O)OC)O)OC(=O)C)CC)OC)C(=O)OC)O.OS(=O)(=O)O
Vincristine sulfate, 99.4%, MP Biomedicals™
CAS: 2068-78-2 Molecular Formula: C46H58N4O14S Molecular Weight (g/mol): 923.044 InChI Key: AQTQHPDCURKLKT-DJOZZVTQSA-N Synonym: vincristine sulfate,methyl 9r,10s,11r,12r,19r-11-acetyloxy-12-ethyl-4-13s,15r,17s-17-ethyl-17-hydroxy-13-methoxycarbonyl-1,11-diazatetracyclo 13.3.1.0?, 1 2 .0?, 1 ? nonadeca-4 12 ,5,7,9-tetraen-13-yl-8-formyl-10-hydroxy-5-methoxy-8,16-diazapentacyclo 10.6.1.0 1 ,?.0 2 ,?.0 PubChem CID: 126963557 SMILES: CCC1(CC2CC(C3=C(CCN(C2)C1)C4=CC=CC=C4N3)(C5=C(C=C6C(=C5)C78CCN9C7C(C=CC9)(C(C(C8N6C=O)(C(=O)OC)O)OC(=O)C)CC)OC)C(=O)OC)O.OS(=O)(=O)O
R&D Systems™ Vincristine sulfate
CAS: 2068-78-2 Molecular Formula: C46H58N4O14S