accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClTiH3CH3COCH3CH3OCH3CH3O

CAS RN 20717-86-6

IUPAC Name: titanium(4+) tris(propan-2-olate) chloride
Synonyms: titanium(4+) tris(propan-2-olate) chloride
Molecular Weight (g/mol): 260.58
Molecular Formula: C9H21ClO3Ti
InChi Key: IFMWVBVPVXRZHE-UHFFFAOYSA-M
SMILES: [Cl-].[Ti+4].CC(C)[O-].CC(C)[O-].CC(C)[O-]
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)