accessibility menu, dialog, popup

UserName

Viewing profile as user:
HO(R)(R)OOHOHOOH

CAS RN 20725-03-5

IUPAC Name: (2R,3R)-2-(3,4-dihydroxyphenyl)-3,7-dihydroxy-3,4-dihydro-2H-1-benzopyran-4-one
Synonyms: tetrahydroxyflavanone dihydrofisetin
Molecular Weight (g/mol): 288.26
Molecular Formula: C15H12O6
InChi Key: FNUPUYFWZXZMIE-LSDHHAIUSA-N
SMILES: O[C@@H]1[C@H](OC2=CC(O)=CC=C2C1=O)C1=CC=C(O)C(O)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results