accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3SOSHONSCH3

CAS RN 207306-50-1

IUPAC Name: 3-[(2-hydroxyethyl)sulfanyl]-6,6-dimethyl-1-(1,3-thiazol-2-yl)-4,5,6,7-tetrahydro-2-benzothiophen-4-one
Synonyms: 3-[(2-hydroxyethyl)sulfanyl]-6,6-dimethyl-1-(1,3-thiazol-2-yl)-5,7-dihydro-2-benzothiophen-4-one
Molecular Weight (g/mol): 339.49
Molecular Formula: C15H17NO2S3
InChi Key: QILRYFCEXLFIDS-UHFFFAOYSA-N
SMILES: CC1(C)CC2=C(SC(SCCO)=C2C(=O)C1)C1=NC=CS1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results