accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3CH3BrCl

CAS RN 2074-80-8

IUPAC Name: 2-bromo-1-chloro-2-methylpropane
Synonyms: 2-bromo-1-chloro-2-methylpropane
Molecular Weight (g/mol): 171.46
Molecular Formula: C4H8BrCl
InChi Key: YEZMNTBSGDUYBT-UHFFFAOYSA-N
SMILES: CC(C)(Br)CCl
Boiling Point 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)