CAS RN 207572-67-6
IUPAC Name:
2-(2-{4-formyl-6-methyl-5-oxo-3-[(phosphonooxy)methyl]-2,5-dihydropyridin-2-ylidene}hydrazin-1-yl)benzene-1,4-disulfonic acid tetrasodium
Synonyms:
2-(2-{4-formyl-6-methyl-5-oxo-3-[(phosphonooxy)methyl]pyridin-2-ylidene}hydrazin-1-yl)benzene-1,4-disulfonic acid tetrasodium
Molecular Weight (g/mol):
603.33
Molecular Formula:
C14H14N3Na4O12PS2
InChi Key:
XPRWSDRXOGVWIR-UHFFFAOYSA-N
SMILES:
[Na].[Na].[Na].[Na].CC1=NC(=NNC2=C(C=CC(=C2)S(O)(=O)=O)S(O)(=O)=O)C(COP(O)(O)=O)=C(C=O)C1=O
1
–
1
of
1
results
1
R&D Systems™ iso-PPADS tetrasodium salt
CAS: 207572-67-6 Molecular Formula: C14H14N3Na4O12PS2