accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CSH

CAS RN 2079-95-0

IUPAC Name: tetradecane-1-thiol
Synonyms: tetradecane-1-thiol
Molecular Weight (g/mol): 230.45
Molecular Formula: C14H30S
InChi Key: GEKDEMKPCKTKEC-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCS
Molecular Weight (g/mol) 
  • (1)
  • (3)
Percent Purity 
  • (1)
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)