Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Tetradecanethiol, 94%

Catalog No. AAL0940114
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0940114 25 g
AAL0940106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0940114 Supplier Thermo Scientific Chemicals Supplier No. L0940114
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 2079-95-0 | C14H30S | 230.454 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2079-95-0
Molecular Formula C14H30S
Molecular Weight (g/mol) 230.454
MDL Number MFCD00022099
InChI Key GEKDEMKPCKTKEC-UHFFFAOYSA-N
Synonym 1-tetradecanethiol, myristyl mercaptan, tetradecanethiol, n-tetradecyl mercaptan, tetradecyl mercaptan, 1-mercaptotetradecane, unii-apy3p7s1xm, apy3p7s1xm, tetradecanethiol-1, acmc-1cq2y
PubChem CID 64677
IUPAC Name tetradecane-1-thiol
SMILES CCCCCCCCCCCCCCS

Specifications

Melting Point 6°C to 7°C
Density 0.85
Boiling Point 176°C to 180°C (22 mmHg)
Flash Point >110°C (230°F)
Odor Unpleasant
Refractive Index 1.461
Linear Formula CH3(CH2)13SH
Quantity 25 g
Beilstein 1742653
Sensitivity Air Sensitive
Formula Weight 230.46
Percent Purity 94%
Chemical Name or Material 1-Tetradecanethiol
Show More Show Less
EINECSNumber 218-209-2
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.