accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COHOH

CAS RN 2081-08-5

IUPAC Name: 4-[1-(4-hydroxyphenyl)ethyl]phenol
Synonyms: 1,1-bis(4-hydroxyphenyl)ethane bisphenol E
Molecular Weight (g/mol): 214.26
Molecular Formula: C14H14O2
InChi Key: HCNHNBLSNVSJTJ-UHFFFAOYSA-N
SMILES: CC(C1=CC=C(O)C=C1)C1=CC=C(O)C=C1
Percent Purity 
  • (1)
  • (1)
Grade 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results