accessibility menu, dialog, popup

UserName

Viewing profile as user:
HNONHNHONNH2

CAS RN 208706-12-1

IUPAC Name: N-{[3-(aminomethyl)cyclohexyl]methyl}-3-(1H-indol-3-yl)-2-[({spiro[indene-1,4'-piperidin]-1'-yl}carbonyl)amino]propanamide
Synonyms: N-{[3-(aminomethyl)cyclohexyl]methyl}-3-(1H-indol-3-yl)-2-({spiro[indene-1,4'-piperidin]-1'-yl}carbonylamino)propanamide
Molecular Weight (g/mol): 539.72
Molecular Formula: C33H41N5O2
InChi Key: DAMXHAMKVXERLM-UHFFFAOYNA-N
SMILES: NCC1CCCC(CNC(=O)C(CC2=CNC3=CC=CC=C23)NC(=O)N2CCC3(CC2)C=CC2=CC=CC=C32)C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results