Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(1R,1'S,3'R/1R,1'R,3'S)-L-054,264, Tocris Bioscience™

Catalog No. 244410
Change view
Click to view available options
Quantity:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
24-441-0 10 mg
24-441 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 24-441-0 Supplier R&D Systems Supplier No. 2444/10

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

This item is not returnable. View return policy

Potent and selective sst2 agonist

Potent and selective somatostatin sst2 receptor agonist (Ki values are 4, 537, 2480, 3614 and 5017 nM for cloned human sst2, sst1, sst4, sst3 and sst5 receptors respectively).

Chemical Identifiers

CAS 208706-12-1
Molecular Formula C33H41N5O2
Molecular Weight (g/mol) 539.724
InChI Key DAMXHAMKVXERLM-DPHZAUTASA-N
Synonym n-2r-1-3-aminomethyl cyclohexyl methyl amino-3-1h-indol-3-yl-1-oxopropan-2-yl spiro indene-1,4'-piperidine-1'-carboxamide
PubChem CID 127256349
IUPAC Name N-[(2R)-1-[[3-(aminomethyl)cyclohexyl]methylamino]-3-(1H-indol-3-yl)-1-oxopropan-2-yl]spiro[indene-1,4'-piperidine]-1'-carboxamide
SMILES C1CC(CC(C1)CNC(=O)C(CC2=CNC3=CC=CC=C32)NC(=O)N4CCC5(CC4)C=CC6=CC=CC=C56)CN

Specifications

Quantity 10 mg
Formula Weight 539.71
Percent Purity >96%
Chemical Name or Material (1R,1'S,3'R/1R,1'R,3'S)-L-054,264
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.