accessibility menu, dialog, popup

UserName

Viewing profile as user:
FSNNNOClClF(?)

CAS RN 209410-46-8

IUPAC Name: 5-(2,6-dichlorophenyl)-2-[(2,4-difluorophenyl)sulfanyl]-6H-pyrimido[1,6-b]pyridazin-6-one
Synonyms: 5-(2,6-dichlorophenyl)-2-[(2,4-difluorophenyl)sulfanyl]pyrimido[1,6-b]pyridazin-6-one
Molecular Weight (g/mol): 436.26
Molecular Formula: C19H9Cl2F2N3OS
InChi Key: VEPKQEUBKLEPRA-UHFFFAOYSA-N
SMILES: FC1=CC=C(SC2=NN3C=NC(=O)C(=C3C=C2)C2=C(Cl)C=CC=C2Cl)C(F)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)