accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 209481-20-9

IUPAC Name: 5-chloro-N-[4-methoxy-3-(piperazin-1-yl)phenyl]-3-methyl-1-benzothiophene-2-sulfonamide hydrochloride
Synonyms: 5-chloro-N-[4-methoxy-3-(piperazin-1-yl)phenyl]-3-methyl-1-benzothiophene-2-sulfonamide hydrochloride
Molecular Weight (g/mol): 488.44
Molecular Formula: C20H23Cl2N3O3S2
InChi Key: RMXZRJYGJMSDQK-UHFFFAOYSA-N
SMILES: Cl.COC1=C(C=C(NS(=O)(=O)C2=C(C)C3=CC(Cl)=CC=C3S2)C=C1)N1CCNCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results