CAS RN 2095432-75-8
IUPAC Name:
N4-[(2H-1,3-benzodioxol-5-yl)methyl]-6-(3-methoxyphenyl)pyrimidine-2,4-diamine hydrochloride
Synonyms:
N4-(2H-1,3-benzodioxol-5-ylmethyl)-6-(3-methoxyphenyl)pyrimidine-2,4-diamine hydrochloride
Molecular Weight (g/mol):
386.84
Molecular Formula:
C19H19ClN4O3
InChi Key:
XZOFNDFDGVAIEH-UHFFFAOYSA-N
SMILES:
Cl.COC1=CC=CC(=C1)C1=CC(NCC2=CC3=C(OCO3)C=C2)=NC(N)=N1
1
–
1
of
1
results
1
R&D Systems™ AMBMP Hydrochloride, >98%
CAS: 2095432-75-8 Molecular Formula: C19H19ClN4O3