CAS RN 209589-59-3
IUPAC Name:
(2S)-5-{[amino(methylsulfanyl)methylidene]amino}-2-azaniumylpentanoate
Synonyms:
(2S)-5-{[amino(methylsulfanyl)methylidene]amino}-2-ammoniopentanoate
Molecular Weight (g/mol):
205.28
Molecular Formula:
C7H15N3O2S
InChi Key:
NGVMVBQRKZPFLB-YFKPBYRVSA-N
SMILES:
CSC(N)=NCCC[C@H]([NH3+])C([O-])=O
1
–
1
of
1
results
1
MilliporeSigma™ Calbiochem™ S-Methyl-L-thiocitrulline, Dihydrochloride
A cell-permeable inhibitor of nitric oxide synthase that exhibits about 17-fold greater selectivity for rat neuronal nitric oxide synthase (IC50 = 300nM) compared to the endothelial enzyme (IC50 = 5.4μM).