accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2096-10-8

IUPAC Name: (2R,3R,4S,5R)-2-(2,6-diamino-9H-purin-9-yl)-5-(hydroxymethyl)oxolane-3,4-diol
Synonyms: 2-aminoadenosine
Molecular Weight (g/mol): 282.26
Molecular Formula: C10H14N6O4
InChi Key: ZDTFMPXQUSBYRL-SBTUYZBDNA-N
SMILES: NC1=NC(N)=C2N=CN([C@@H]3O[C@H](CO)[C@@H](O)[C@H]3O)C2=N1
Molecular Weight (g/mol) 
  • (1)
  • (5)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)
Physical Form 
  • (2)
Solvent or Matrix 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (3)
  • (1)
  • (5)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)