Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Aminoadenosine 98.0+%, TCI America™
SDP

Catalog No. A21355G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A21355G 5 g
A213525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A21355G Supplier TCI America Supplier No. A21355G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 2096-10-8
Molecular Formula C10H14N6O4
Molecular Weight (g/mol) 282.26
MDL Number MFCD00053556
InChI Key ZDTFMPXQUSBYRL-SBTUYZBDNA-N
Synonym 2-aminoadenosine, 2-amine adenosine, adenosine, 2-amino, 2,6-diaminopurine riboside, 2,6-diaminonebularine, 2,6-diaminopurinosine, 2r,3r,4s,5r-2-2,6-diaminopurin-9-yl-5-hydroxymethyl tetrahydrofuran-3,4-diol, 2,6-diamino-9-beta-d-ribofuranosyl purine, pubchem14156, ksc202m1p
PubChem CID 72200
ChEBI CHEBI:1014
IUPAC Name (2R,3R,4S,5R)-2-(2,6-diamino-9H-purin-9-yl)-5-(hydroxymethyl)oxolane-3,4-diol
SMILES NC1=NC(N)=C2N=CN([C@@H]3O[C@H](CO)[C@@H](O)[C@H]3O)C2=N1

Specifications

Melting Point 252°C
Color White-Yellow
Quantity 5 g
Formula Weight 282.26
Percent Purity ≥98.0% (HPLC,T)
Grade Not Available
Physical Form Crystalline Powder
Chemical Name or Material 2-Aminoadenosine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.