accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2096330-52-6

IUPAC Name: {3-[methyl(phenyl)sulfamoyl]phenyl}boronic acid
Synonyms: 3-[methyl(phenyl)sulfamoyl]phenylboronic acid
Molecular Weight (g/mol): 291.13
Molecular Formula: C13H14BNO4S
InChi Key: LQZLHQGOBLQKOM-UHFFFAOYSA-N
SMILES: CN(C1=CC=CC=C1)S(=O)(=O)C1=CC=CC(=C1)B(O)O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results