accessibility menu, dialog, popup

UserName

Viewing profile as user:
BONHFNOOClHOOH

CAS RN 2096332-75-9

IUPAC Name: {2-chloro-5-[(4-fluoro-2-nitrophenyl)carbamoyl]phenyl}boronic acid
Synonyms: 2-chloro-5-[(4-fluoro-2-nitrophenyl)carbamoyl]phenylboronic acid
Molecular Weight (g/mol): 338.48
Molecular Formula: C13H9BClFN2O5
InChi Key: HBBVKWQZDSTEMK-UHFFFAOYSA-N
SMILES: OB(O)C1=C(Cl)C=CC(=C1)C(=O)NC1=C(C=C(F)C=C1)[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results