CAS RN 2096332-79-3
IUPAC Name:
(5-{[2-(ethylamino)ethyl]carbamoyl}-2-fluorophenyl)boronic acid
Synonyms:
5-{[2-(ethylamino)ethyl]carbamoyl}-2-fluorophenylboronic acid
Molecular Weight (g/mol):
254.07
Molecular Formula:
C11H16BFN2O3
InChi Key:
UBDRCBYERBTPRG-UHFFFAOYSA-N
SMILES:
CCNCCNC(=O)C1=CC(B(O)O)=C(F)C=C1
1
–
1
of
1
results
1
5-[2-(Ethylamino)ethylcarbamoyl]-2-fluorobenzeneboronic acid, 97%, Thermo Scientific™
CAS: 2096332-79-3 Molecular Formula: C11H16BFN2O3 Molecular Weight (g/mol): 254.07 MDL Number: MFCD20040204 InChI Key: UBDRCBYERBTPRG-UHFFFAOYSA-N Synonym: 5-2-ethylamino ethylcarbamoyl-2-fluorobenzeneboronic acid PubChem CID: 73995842