accessibility menu, dialog, popup

UserName

Viewing profile as user:
BOHNFClClHOOH

CAS RN 2096333-19-4

IUPAC Name: {2-chloro-5-[(3-chloro-4-fluorophenyl)carbamoyl]phenyl}boronic acid
Synonyms: 2-chloro-5-[(3-chloro-4-fluorophenyl)carbamoyl]phenylboronic acid
Molecular Weight (g/mol): 327.93
Molecular Formula: C13H9BCl2FNO3
InChi Key: SGQGAHDHVCIEAK-UHFFFAOYSA-N
SMILES: OB(O)C1=C(Cl)C=CC(=C1)C(=O)NC1=CC(Cl)=C(F)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results