CAS RN 2096333-34-3
IUPAC Name:
(2-fluoro-5-{[(2-fluoro-5-methylphenyl)methyl]carbamoyl}phenyl)boronic acid
Synonyms:
2-fluoro-5-{[(2-fluoro-5-methylphenyl)methyl]carbamoyl}phenylboronic acid
Molecular Weight (g/mol):
305.09
Molecular Formula:
C15H14BF2NO3
InChi Key:
BBGVRRJZNLKTKZ-UHFFFAOYSA-N
SMILES:
CC1=CC(CNC(=O)C2=CC(B(O)O)=C(F)C=C2)=C(F)C=C1
1
–
1
of
1
results
1
2-Fluoro-5-(2-fluoro-5-methylbenzylcarbamoyl)benzeneboronic acid, 97%, Thermo Scientific™
CAS: 2096333-34-3 Molecular Formula: C15H14BF2NO3 Molecular Weight (g/mol): 305.09 MDL Number: MFCD20091226 InChI Key: BBGVRRJZNLKTKZ-UHFFFAOYSA-N Synonym: 2-fluoro-5-2-fluoro-5-methylbenzylcarbamoyl benzeneboronic acid PubChem CID: 73996013