accessibility menu, dialog, popup

UserName

Viewing profile as user:
BSNNHOOH

CAS RN 2096334-52-8

IUPAC Name: {4-[2-(5-phenylpyridin-2-yl)-1,3-thiazol-4-yl]phenyl}boronic acid
Synonyms: 4-[2-(5-phenylpyridin-2-yl)-1,3-thiazol-4-yl]phenylboronic acid
Molecular Weight (g/mol): 358.22
Molecular Formula: C20H15BN2O2S
InChi Key: BSXIBVIMFORFRB-UHFFFAOYSA-N
SMILES: OB(O)C1=CC=C(C=C1)C1=CSC(=N1)C1=NC=C(C=C1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results