accessibility menu, dialog, popup

UserName

Viewing profile as user:
BFOHNCH3HOOH

CAS RN 2096337-33-4

IUPAC Name: {4-fluoro-3-[(3-methylphenyl)carbamoyl]phenyl}boronic acid
Synonyms: 4-fluoro-3-[(3-methylphenyl)carbamoyl]phenylboronic acid
Molecular Weight (g/mol): 273.07
Molecular Formula: C14H13BFNO3
InChi Key: KAEODVFXXIOTPP-UHFFFAOYSA-N
SMILES: CC1=CC(NC(=O)C2=C(F)C=CC(=C2)B(O)O)=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results