accessibility menu, dialog, popup

UserName

Viewing profile as user:
BSNNNHOOH

CAS RN 2096338-84-8

IUPAC Name: {4'-[2-(pyrazin-2-yl)-1,3-thiazol-4-yl]-[1,1'-biphenyl]-4-yl}boronic acid
Synonyms: 4'-[2-(pyrazin-2-yl)-1,3-thiazol-4-yl]-[1,1'-biphenyl]-4-ylboronic acid
Molecular Weight (g/mol): 359.21
Molecular Formula: C19H14BN3O2S
InChi Key: QIJNANKKYORYSQ-UHFFFAOYSA-N
SMILES: OB(O)C1=CC=C(C=C1)C1=CC=C(C=C1)C1=CSC(=N1)C1=NC=CN=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results