CAS RN 2098799-78-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Tocris Bioscience™ VH 032, propargyl
Alkyne-functionalized VH 032; 'click' VHL ligand for PROTAC™s