accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 209991-64-0

IUPAC Name: 2-[2-fluoro-4-(trifluoromethyl)phenyl]acetic acid
Synonyms: [2-fluoro-4-(trifluoromethyl)phenyl]acetic acid
Molecular Weight (g/mol): 222.14
Molecular Formula: C9H6F4O2
InChi Key: LUIOBGWYVHPKRC-UHFFFAOYSA-N
SMILES: OC(=O)CC1=C(F)C=C(C=C1)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results