CAS RN 210110-90-0
IUPAC Name:
1-butyl-2-(hydroxymethyl)piperidine-3,4,5-triol hydrochloride
Synonyms:
1-butyl-2-(hydroxymethyl)piperidine-3,4,5-triol hydrochloride
Molecular Weight (g/mol):
255.74
Molecular Formula:
C10H22ClNO4
InChi Key:
QPAFAUYWVZMWPR-UHFFFAOYNA-N
SMILES:
Cl.CCCCN1CC(O)C(O)C(O)C1CO
1
–
1
of
1
results
1
R&D Systems™ Miglustat hydrochloride
CAS: 210110-90-0 Molecular Formula: C10H22ClNO4