accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 21043-40-3

NHH2N

CAS RN 21043-40-3

IUPAC Name: 4-cyclopentylpiperazine-1,4-diium
Synonyms: 4-cyclopentylpiperazine-1,4-diium
Molecular Weight (g/mol): 156.27
Molecular Formula: C9H20N2
InChi Key: PVMCQBPJKPMOKM-UHFFFAOYSA-P
SMILES: C1CCC(C1)[NH+]1CC[NH2+]CC1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results