CAS RN 210688-56-5
IUPAC Name:
3-{2-[4-(3,4-dimethylphenyl)piperazin-1-yl]ethyl}-2,3-dihydro-1H-isoindol-1-one dihydrochloride
Synonyms:
3-{2-[4-(3,4-dimethylphenyl)piperazin-1-yl]ethyl}-2,3-dihydroisoindol-1-one dihydrochloride
Molecular Weight (g/mol):
422.39
Molecular Formula:
C22H29Cl2N3O
InChi Key:
RMNWLEGGYCVHFD-UHFFFAOYNA-N
SMILES:
Cl.Cl.CC1=C(C)C=C(C=C1)N1CCN(CCC2NC(=O)C3=CC=CC=C23)CC1
1
–
1
of
1
results
1
R&D Systems™ PD 168568 dihydrochloride
CAS: 210688-56-5 Molecular Formula: C22H29Cl2N3O