CAS RN 210880-92-5
CAS RN 210880-92-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
TraceCERT™ Clothianidin, MilliporeSigma™ Supelco™
Find more information here: Neonicotinoids