accessibility menu, dialog, popup

UserName

Viewing profile as user:
NHONHNHNNNBr

CAS RN 211096-49-0

IUPAC Name: 1-(2-bromophenyl)-3-(7-cyano-2H-1,2,3-benzotriazol-4-yl)urea
Synonyms: 1-(2-bromophenyl)-3-(7-cyano-2H-1,2,3-benzotriazol-4-yl)urea
Molecular Weight (g/mol): 357.17
Molecular Formula: C14H9BrN6O
InChi Key: SEDUMQWZEOMXSO-UHFFFAOYSA-N
SMILES: BrC1=CC=CC=C1NC(=O)NC1=CC=C(C#N)C2=NNN=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (3)