accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3CH3OOHOH3CHO

CAS RN 211110-63-3

IUPAC Name: 2-(4-{[4-hydroxy-3-(propan-2-yl)phenyl]methyl}-3,5-dimethylphenoxy)acetic acid
Synonyms: 4-[(4-hydroxy-3-isopropylphenyl)methyl]-3,5-dimethylphenoxyacetic acid
Molecular Weight (g/mol): 328.41
Molecular Formula: C20H24O4
InChi Key: QNAZTOHXCZPOSA-UHFFFAOYSA-N
SMILES: CC(C)C1=CC(CC2=C(C)C=C(OCC(O)=O)C=C2C)=CC=C1O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)