CAS RN 21149-17-7
CAS RN 21149-17-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Promotions Available
Sigma Aldrich 3-(7-(Tert-Butoxycarbonyl)-5 6 7 8-Tetrahydroimidazo[1 2-A]Pyrazin-3-Yl)Propanoic Acid
Sigma Aldrich 3-(7-(Tert-Butoxycarbonyl)-5 6 7 8-Tetrahydroimidazo[1 2-A]Pyrazin-3-Yl)Propanoic Acid
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.