CAS RN 211512-13-9
CAS RN 211512-13-9
IUPAC Name:
N-(4-aminobutyl)-2-nitrobenzene-1-sulfonamide
Synonyms:
N-(4-aminobutyl)-2-nitrobenzenesulfonamide
Molecular Weight (g/mol):
273.31
Molecular Formula:
C10H15N3O4S
InChi Key:
NEIRGPMZEKSCSV-UHFFFAOYSA-N
SMILES:
NCCCCNS(=O)(=O)C1=CC=CC=C1[N+]([O-])=O
1
–
1
of
1
results
1
N-(4-Aminobutyl)-2-nitrobenzenesulfonamide 95.0+%, TCI America™
CAS: 211512-13-9 Molecular Formula: C10H15N3O4S Molecular Weight (g/mol): 273.307 MDL Number: MFCD04117909 InChI Key: NEIRGPMZEKSCSV-UHFFFAOYSA-N Synonym: 1-Amino-4-(2-nitrobenzenesulfonamido)butane, N-(2-Nitrobenzenesulfonyl)-1,4-butanediamine, N-(o-Ns)-1,4-diaminobutane PubChem CID: 11323272 IUPAC Name: N-(4-aminobutyl)-2-nitrobenzenesulfonamide SMILES: C1=CC=C(C(=C1)[N+](=O)[O-])S(=O)(=O)NCCCCN