CAS RN 2122196-16-9
IUPAC Name:
4-[2-({6-amino-9-[3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-9H-purin-2-yl}sulfanyl)ethyl]benzene-1-sulfonic acid amine
Synonyms:
4-[2-({6-amino-9-[3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]purin-2-yl}sulfanyl)ethyl]benzenesulfonic acid amine
Molecular Weight (g/mol):
500.55
Molecular Formula:
C18H24N6O7S2
InChi Key:
AYQGFWKUGKSGCZ-UHFFFAOYNA-N
SMILES:
N.NC1=C2N=CN(C3OC(CO)C(O)C3O)C2=NC(SCCC2=CC=C(C=C2)S(O)(=O)=O)=N1
1
–
1
of
1
results
1
R&D Systems™ PSB 0777 ammonium salt
CAS: 2122196-16-9 Molecular Formula: C18H24N6O7S2