accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 212250-92-5

IUPAC Name: 2-{[(2-{[3-oxo-2-(2,4,6-trimethylbenzoyl)but-1-en-1-yl]amino}-1,2-diphenylethyl)amino]methylidene}-1-(2,4,6-trimethylphenyl)butane-1,3-dione cobalt
Synonyms: 2-{[(2-{[3-oxo-2-(2,4,6-trimethylbenzoyl)but-1-en-1-yl]amino}-1,2-diphenylethyl)amino]methylidene}-1-(2,4,6-trimethylphenyl)butane-1,3-dione cobalt
Molecular Weight (g/mol): 699.76
Molecular Formula: C42H44CoN2O4
InChi Key: KKCIKZXUKHOLHL-UHFFFAOYNA-N
SMILES: [Co].CC(=O)C(=CNC(C(NC=C(C(C)=O)C(=O)C1=C(C)C=C(C)C=C1C)C1=CC=CC=C1)C1=CC=CC=C1)C(=O)C1=C(C)C=C(C)C=C1C
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results