CAS RN 212391-63-4
IUPAC Name:
6-(2,6-dichlorophenyl)-2-({4-[2-(diethylamino)ethoxy]phenyl}amino)-8-methyl-7H,8H-pyrido[2,3-d]pyrimidin-7-one dihydrochloride
Synonyms:
6-(2,6-dichlorophenyl)-2-({4-[2-(diethylamino)ethoxy]phenyl}amino)-8-methylpyrido[2,3-d]pyrimidin-7-one dihydrochloride
Molecular Weight (g/mol):
585.35
Molecular Formula:
C26H29Cl4N5O2
InChi Key:
NADLBPWBFGTESN-UHFFFAOYSA-N
SMILES:
Cl.Cl.CCN(CC)CCOC1=CC=C(NC2=NC=C3C=C(C(=O)N(C)C3=N2)C2=C(Cl)C=CC=C2Cl)C=C1
1
–
1
of
1
results
1
R&D Systems™ PD 166285 dihydrochloride
CAS: 212391-63-4 Molecular Formula: C26H29Cl4N5O2