accessibility menu, dialog, popup

UserName

Viewing profile as user:
ONHOFFHNICl

CAS RN 212631-79-3

IUPAC Name: 2-[(2-chloro-4-iodophenyl)amino]-N-(cyclopropylmethoxy)-3,4-difluorobenzamide
Synonyms: 2-[(2-chloro-4-iodophenyl)amino]-N-(cyclopropylmethoxy)-3,4-difluorobenzamide
Molecular Weight (g/mol): 478.66
Molecular Formula: C17H14ClF2IN2O2
InChi Key: GFMMXOIFOQCCGU-UHFFFAOYSA-N
SMILES: FC1=C(F)C(NC2=C(Cl)C=C(I)C=C2)=C(C=C1)C(=O)NOCC1CC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results