accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 213627-19-1

IUPAC Name: magnesium(2+) (2,5-dichlorophenyl)methanide chloride
Synonyms: magnesium(2+) (2,5-dichlorophenyl)methanide chloride
Molecular Weight (g/mol): 219.77
Molecular Formula: C7H5Cl3Mg
InChi Key: LCSTXMZCRKYVRE-UHFFFAOYSA-M
SMILES: [Mg++].[Cl-].[CH2-]C1=C(Cl)C=CC(Cl)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results