CAS RN 2137047-43-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Tocris Bioscience™ VU 6005649
Positive allosteric modulators of mGlu7 and mGlu8 receptors. Also NK1 antagonist.