accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHNHOBrBr(E/Z)

CAS RN 21386-43-6

IUPAC Name: 2,4-dibromo-6-[(hydroxyamino)methylidene]cyclohexa-2,4-dien-1-one
Synonyms: 2,4-dibromo-6-[(hydroxyamino)methylidene]cyclohexa-2,4-dien-1-one
Molecular Weight (g/mol): 294.93
Molecular Formula: C7H5Br2NO2
InChi Key: LRYPGXXEEAWIBE-UHFFFAOYSA-N
SMILES: ONC=C1C=C(Br)C=C(Br)C1=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results