accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NSS

CAS RN 21402-19-7

IUPAC Name: 3-methyl-4-phenyl-2,3-dihydro-1,3-thiazole-2-thione
Synonyms: 3-methyl-4-phenyl-1,3-thiazole-2-thione
Molecular Weight (g/mol): 207.31
Molecular Formula: C10H9NS2
InChi Key: YGZDFPNBMUHUON-UHFFFAOYSA-N
SMILES: CN1C(=S)SC=C1C1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results