Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Methyl-4-phenylthiazoline-2-thione 98.0+%, TCI America™
SDP

Catalog No. M03551G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M03551G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M03551G Supplier TCI America Supplier No. M03551G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 21402-19-7
Molecular Formula C10H9NS2
Molecular Weight (g/mol) 207.31
MDL Number MFCD00059731
InChI Key YGZDFPNBMUHUON-UHFFFAOYSA-N
PubChem CID 519864
IUPAC Name 3-methyl-4-phenyl-2,3-dihydro-1,3-thiazole-2-thione
SMILES CN1C(=S)SC=C1C1=CC=CC=C1

Specifications

Melting Point 130°C
Color Yellow
Quantity 1 g
Formula Weight 207.31
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Methyl-4-phenylthiazoline-2-thione
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.