accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CON

CAS RN 2141-62-0

IUPAC Name: 3-ethoxypropanenitrile
Synonyms: 3-ethoxy-propionitrile
Molecular Weight (g/mol): 99.13
Molecular Formula: C5H9NO
InChi Key: DCWQZPJHHVLHSV-UHFFFAOYSA-N
SMILES: CCOCCC#N
Boiling Point 
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)