Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Ethoxypropionitrile, 95%, Thermo Scientific™

Catalog No. AAA14243AP
Change view
Click to view available options
Quantity:
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA14243AP 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAA14243AP Supplier Thermo Scientific Chemicals Supplier No. A14243AP

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 2141-62-0
Boiling Point 171°C to 173°C
Flash Point 63°C (145°F)
Molecular Formula C5H9NO
Linear Formula CH3CH2OCH2CH2CN
Quantity 500mL
MDL Number MFCD00001959
Beilstein 741924
Synonym 3-ethoxypropionitrile, propanenitrile, 3-ethoxy, propionitrile, 3-ethoxy, 3-ethoxy-propionitrile, 3-ethoxypropiononitrile, beta-ethoxypropionitrile, 2-ethoxypropionitrile, alpha-ethoxypropionitrile, propionitrile, 2-ethoxy, .beta.-ethoxypropionitrile
InChI Key DCWQZPJHHVLHSV-UHFFFAOYSA-N
SMILES CCOCCC#N
IUPAC Name 3-ethoxypropanenitrile
Molecular Weight (g/mol) 99.133
PubChem CID 16501
Formula Weight 99.14
Percent Purity 95%
Chemical Name or Material 3-Ethoxypropionitrile
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Precautionary Statement GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
RTECSNumber TZ4680000
TSCA TSCA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.