accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOHH3C(E/Z)

CAS RN 215030-90-3

IUPAC Name: 4-{2-[5,5-dimethyl-8-(2-phenylethynyl)-5,6-dihydronaphthalen-2-yl]ethenyl}benzoic acid
Synonyms: 4-{2-[5,5-dimethyl-8-(2-phenylethynyl)-6H-naphthalen-2-yl]ethenyl}benzoic acid
Molecular Weight (g/mol): 404.51
Molecular Formula: C29H24O2
InChi Key: YCADIXLLWMXYKW-UHFFFAOYSA-N
SMILES: CC1(C)CC=C(C#CC2=CC=CC=C2)C2=C1C=CC(C=CC1=CC=C(C=C1)C(O)=O)=C2

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)