accessibility menu, dialog, popup

UserName

Viewing profile as user:
HO(R)OHOH(R)OHOH

CAS RN 2152-56-9

IUPAC Name: (2R,4R)-pentane-1,2,3,4,5-pentol
Synonyms: arabitol D-arabinitol
Molecular Weight (g/mol): 152.15
Molecular Formula: C5H12O5
InChi Key: HEBKCHPVOIAQTA-QWWZWVQMSA-N
SMILES: OC[C@@H](O)C(O)[C@H](O)CO
Grade 
  • (1)
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)