Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DL-Arabitol 97.0+%, TCI America™
SDP

Catalog No. A0517100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A0517100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A0517100MG Supplier TCI America Supplier No. A0517100MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

High-purity laboratory reagent for research applications.

Chemical Identifiers

CAS 2152-56-9
Molecular Formula C5H12O5
Molecular Weight (g/mol) 152.146
MDL Number MFCD00070503
InChI Key HEBKCHPVOIAQTA-QWWZWVQMSA-N
Synonym DL-Arabinitol
PubChem CID 94154
ChEBI CHEBI:18333
IUPAC Name (2R,4R)-pentane-1,2,3,4,5-pentol
SMILES C(C(C(C(CO)O)O)O)O

Specifications

Melting Point 106°C
Color White
Quantity 100 mg
Formula Weight 152.15
Percent Purity ≥97.0% (GC)
Grade Not Available
Physical Form Crystalline Powder
Chemical Name or Material DL-Arabitol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.